4-(Dimethylcarbamoyl)-2-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Dimethylcarbamoyl-2-fluorobenzeneboronic acid
BB-3325-5G
BB-3325-1G
BB-3325-250M
COBBBB-3325-5GEA
0
EUR
COBBBB-3325-5G
COBBBB-3325-1G
COBBBB-3325-250M
4-(Dimethylcarbamoyl)-2-fluorphenylboronsäure
4-(Dimethylcarbamoyl)-2-fluorphenylboronsäure
Formel:
C₉H₁₁BFNO₃ |
MDL:
MFCD11855833 CAS-Nummer: 874289-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...