4-(Dimethylcarbamoyl)-3-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Dimethylcarbamoyl-3-fluorobenzeneboronic acid
BB-3309-1G
BB-3309-5G
COBBBB-3309-1GEA
301
EUR
COBBBB-3309-1G
COBBBB-3309-5G
4-(Dimethylcarbamoyl)-3-fluorphenylboronsäure
4-(Dimethylcarbamoyl)-3-fluorphenylboronsäure
Formel:
C₉H₁₁BFNO₃ Molecular Weight: 211.00 g/mol |
MDL:
MFCD08436060 CAS-Nummer: 874289-13-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...