3-Fluor-4-(methylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Fluoro-4-(methylcarbamoyl)benzeneboronic acid, (3-Fluoro-4-(methylcarbamoyl)phenyl)boronic acid

BB-3303-5G BB-3303-1G
COBBBB-3303-5GEA 1310 EUR
COBBBB-3303-5G COBBBB-3303-1G
3-Fluor-4-(methylcarbamoyl)phenylboronsäure
3-Fluor-4-(methylcarbamoyl)phenylboronsäure
Formel: C₈H₉BFNO₃
Molecular Weight: 196.97 g/mol
MDL: MFCD08436026
CAS-Nummer: 849833-86-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR