5-Fluor-2-(methylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-Fluoro-2-(methylcarbamoyl)phenyl)boronic acid

BB-5093-5G BB-5093-1G
COBBBB-5093-5GEA 0 EUR
COBBBB-5093-5G COBBBB-5093-1G
5-Fluor-2-(methylcarbamoyl)phenylboronsäure
5-Fluor-2-(methylcarbamoyl)phenylboronsäure
Formel: C₈H₉BFNO₃
MDL: MFCD09878343
CAS-Nummer: 957062-96-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR