3-Fluor-5-(methylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Fluoro-5-(methylcarbamoyl)phenyl)boronic acid, 3-Fluoro-5-(methylcarbamoyl)benzeneboronic acid

BB-2802-5G BB-2802-1G
COBBBB-2802-5GEA 0 EUR
COBBBB-2802-5G COBBBB-2802-1G
3-Fluor-5-(methylcarbamoyl)phenylboronsäure
3-Fluor-5-(methylcarbamoyl)phenylboronsäure
Formel: C₈H₉BFNO₃
MDL: MFCD07363793
CAS-Nummer: 871332-63-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR