4-(Ethoxycarbonyl)-3-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Ethoxycarbonyl-3-fluorobenzeneboronic acid, 4-Ethoxycarbonyl-3-fluorophenylboronic acid

BB-3301-5G BB-3301-1G
COBBBB-3301-5GEA 507 EUR
COBBBB-3301-5G COBBBB-3301-1G
4-(Ethoxycarbonyl)-3-fluorphenylboronsäure
4-(Ethoxycarbonyl)-3-fluorphenylboronsäure
Formel: C₉H₁₀BFO₄
Molecular Weight: 211.98 g/mol
MDL: MFCD06656274
CAS-Nummer: 874288-38-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR