4-(Ethoxycarbonyl)-3-fluorphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 4-Ethoxycarbonyl-3-fluorobenzeneboronic acid, 4-Ethoxycarbonyl-3-fluorophenylboronic acid

016209-1G 016209-5G
FLUO016209-1GEA 155 EUR
FLUO016209-1G FLUO016209-5G
4-(Ethoxycarbonyl)-3-fluorphenylboronsäure
4-(Ethoxycarbonyl)-3-fluorphenylboronsäure
Formel: C₉H₁₀BFO₄
Molecular Weight: 211.98 g/mol
MDL: MFCD06656274
CAS-Nummer: 874288-38-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR