3-Fluor-(4-methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Fluoro-4-(methoxycarbonyl)phenyl)boronic acid, 3-Fluoro-4-(methoxycarbonyl)phenylboronic acid, 3-Fluoro-4-methoxycarbonylphenylboronic acid, 3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid

BB-2719-1G BB-2719-25G BB-2719-5G
COBBBB-2719-1GEA 0 EUR
COBBBB-2719-1G COBBBB-2719-25G COBBBB-2719-5G
3-Fluor-(4-methoxycarbonyl)phenylboronsäure
3-Fluor-(4-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BFO₄
MDL: MFCD04115665
CAS-Nummer: 505083-04-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR