2-(Ethoxycarbonyl)-4-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Ethoxycarbonyl)-4-fluorophenyl)boronic acid

BB-5295-5G BB-5295-1G
COBBBB-5295-5GEA 1020 EUR
COBBBB-5295-5G COBBBB-5295-1G
2-(Ethoxycarbonyl)-4-fluorphenylboronsäure
2-(Ethoxycarbonyl)-4-fluorphenylboronsäure
Formel: C₉H₁₀BFO₄
Molecular Weight: 211,9800 g/mol
MDL: MFCD11053864
CAS-Nummer: 1072951-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR