4-Fluor-(3-methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid, 4-Fluoro-3-methoxycarbonylphenylboronic acid

BB-2860-1G BB-2860-25G BB-2860-5G
COBBBB-2860-1GEA 0 EUR
COBBBB-2860-1G COBBBB-2860-25G COBBBB-2860-5G
4-Fluor-(3-methoxycarbonyl)phenylboronsäure
4-Fluor-(3-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BFO₄
Molecular Weight: 197.96 g/mol
MDL: MFCD06801688
CAS-Nummer: 874219-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR