2-(Ethoxycarbonyl)-5-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Ethoxycabonyl-5-fluorophenylboronic acid, (2-(Ethoxycarbonyl)-5-fluorophenyl)boronic acid
BB-5088-5G
BB-5088-1G
COBBBB-5088-5GEA
834
EUR
COBBBB-5088-5G
COBBBB-5088-1G
2-(Ethoxycarbonyl)-5-fluorphenylboronsäure
2-(Ethoxycarbonyl)-5-fluorphenylboronsäure
Formel:
C₉H₁₀BFO₄ Molecular Weight: 211.9800 g/mol |
MDL:
MFCD09878342 CAS-Nummer: 957062-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...