5-Fluor-(2-methoxycarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
5-Fluoro-2-methoxycarboxyphenylboronic acid, 5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid, 2-Methoxycarbonyl-5-fluorophenylboronic acid
BB-2727-5G
BB-2727-1G
COBBBB-2727-5GEA
0
EUR
COBBBB-2727-5G
COBBBB-2727-1G
5-Fluor-(2-methoxycarbonyl)phenylboronsäure
5-Fluor-(2-methoxycarbonyl)phenylboronsäure
Formel:
C₈H₈BFO₄ Molecular Weight: 197.96 g/mol |
MDL:
MFCD03095367 CAS-Nummer: 850568-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...