3-(Ethoxycarbonyl)-5-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-(Ethoxycarbonyl)-5-fluorophenyl)boronic acid, 3-Fluoro-5-(ethoxycarbonyl)benzeneboronic acid
BB-2800-5G
BB-2800-1G
COBBBB-2800-5GEA
1740
EUR
COBBBB-2800-5G
COBBBB-2800-1G
3-(Ethoxycarbonyl)-5-fluorphenylboronsäure
3-(Ethoxycarbonyl)-5-fluorphenylboronsäure
Formel:
C₉H₁₀BFO₄ Molecular Weight: 211,9800 g/mol |
MDL:
MFCD07363792 CAS-Nummer: 871329-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...