3-Fluor-(5-methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Fluoro-5-methoxycarbonylphenylboronic Acid, 3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid

BB-2355-1G BB-2355-5G
COBBBB-2355-1GEA 0 EUR
COBBBB-2355-1G COBBBB-2355-5G
3-Fluor-(5-methoxycarbonyl)phenylboronsäure
3-Fluor-(5-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BFO₄
Molecular Weight: 197.96 g/mol
MDL: MFCD07363749
CAS-Nummer: 871329-62-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR