2-Fluor-(5-methoxycarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Fluoro-5-(methoxycarbonyl)phenylboronic acid, (2-Fluoro-5-(methoxycarbonyl)phenyl)boronic acid
BB-2726-1G
BB-2726-5G
COBBBB-2726-1GEA
98.7
EUR
COBBBB-2726-1G
COBBBB-2726-5G
2-Fluor-(5-methoxycarbonyl)phenylboronsäure
2-Fluor-(5-methoxycarbonyl)phenylboronsäure
Formel:
C₈H₈BFO₄ Molecular Weight: 197.9600 g/mol |
MDL:
MFCD06659868 CAS-Nummer: 850568-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...