3-Carboxy-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Carboxy-2-fluorobenzeneboronic acid, 3-Borono-2-fluorobenzoic acid

BB-8386-5G BB-8386-1G
COBBBB-8386-5GEA 114 EUR
COBBBB-8386-5G COBBBB-8386-1G
3-Carboxy-2-fluorphenylboronsäure
3-Carboxy-2-fluorphenylboronsäure
Formel: C₇H₆BFO₄
Molecular Weight: 183,9300 g/mol
MDL: MFCD03095128
CAS-Nummer: 1072952-09-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR