4-Carboxy-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Carboxy-2-fluorobenzeneboronic acid

BB-3230-5G BB-3230-25G BB-3230-1G
COBBBB-3230-5GEA 359 EUR
COBBBB-3230-5G COBBBB-3230-25G COBBBB-3230-1G
4-Carboxy-2-fluorphenylboronsäure
4-Carboxy-2-fluorphenylboronsäure
Formel: C₇H₆BFO₄
Molecular Weight: 183,93 g/mol
MDL: MFCD08458473
CAS-Nummer: 851135-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR