4-Carboxy-3-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Borono-2-fluorobenzoic acid, 4-Carboxy-3-fluorobenzeneboronic acid

BB-2632-25G BB-2632-5G
COBBBB-2632-25GEA 314 EUR
COBBBB-2632-25G COBBBB-2632-5G
4-Carboxy-3-fluorphenylboronsäure
4-Carboxy-3-fluorphenylboronsäure
Formel: C₇H₆BFO₄
Molecular Weight: 183,93 g/mol
MDL: MFCD01114671
CAS-Nummer: 120153-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR