2-Carboxy-5-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Carboxy-5-flourophenylboronic acid
BB-3934-1G
BB-3934-25G
BB-3934-5G
COBBBB-3934-1GEA
75.9
EUR
COBBBB-3934-1G
COBBBB-3934-25G
COBBBB-3934-5G
2-Carboxy-5-fluorphenylboronsäure
2-Carboxy-5-fluorphenylboronsäure
Formel:
C₇H₆BFO₄ |
MDL:
MFCD06656266 CAS-Nummer: 874290-62-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...