3-Carboxy-5-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Carboxy-5-fluorobenzeneboronic acid, 3-Borono-5-fluorobenzoic acid

BB-2797-5G BB-2797-1G
COBBBB-2797-5GEA 303 EUR
COBBBB-2797-5G COBBBB-2797-1G
3-Carboxy-5-fluorphenylboronsäure
3-Carboxy-5-fluorphenylboronsäure
Formel: C₇H₆BFO₄
Molecular Weight: 183.93 g/mol
MDL: MFCD07363791
CAS-Nummer: 871329-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR