N,N-Dimethyl-4-fluor-2-nitroanilin
Lieferant: CHEMPUR
Synonyme:
4-Fluoro-N,N-dimethyl-2-nitroaniline, N,N-Dimethyl 4-fluoro-2-nitroaniline
FL13312.POR
013312-1G
013312-5G
CHMPFL13312.POREA
0
EUR
CHMPFL13312.POR
FLUO013312-1G
FLUO013312-5G
N,N-Dimethyl-4-fluor-2-nitroanilin
N,N-Dimethyl-4-fluor-2-nitroanilin
Formel:
C₈H₉FN₂O₂ Molecular Weight: 184,17 g/mol |
MDL:
MFCD04116334 CAS-Nummer: 69261-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...