N-tert-Butyl-5-fluor-2-nitroanilin
Lieferant: COMBI-BLOCKS
Synonyme:
N-(tert-Butyl)-5-fluoro-2-nitroaniline
AN-1421-5G
AN-1421-1G
COBBAN-1421-5GEA
451
EUR
COBBAN-1421-5G
COBBAN-1421-1G
N-tert-Butyl-5-fluor-2-nitroanilin
N-tert-Butyl-5-fluor-2-nitroanilin
Formel:
C₁₀H₁₃FN₂O₂ Molecular Weight: 212,220 g/mol |
MDL:
MFCD11855826 CAS-Nummer: 1133115-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...