N-Methyl-2-nitro-4-(trifluormethoxy)anilin

Lieferant: COMBI-BLOCKS

HC-3320-1G HC-3320-5G
COBBHC-3320-1GEA 303 EUR
COBBHC-3320-1G COBBHC-3320-5G
N-Methyl-2-nitro-4-(trifluormethoxy)anilin
N-Methyl-2-nitro-4-(trifluormethoxy)anilin
Formel: C₈H₇F₃N₂O₃
MDL: MFCD15143571
CAS-Nummer: 1215206-37-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR