Anilinsulfat ≥98%
Lieferant: Thermo Fisher Scientific
Gefahr
A15563.22 A15563.36
A15563.22EA 28.2 EUR
A15563.22 A15563.36
Anilinsulfat ≥98%
Anilinsulfat
Purity: 98% CAS No.: 542-16-5 Molecular Formula: (C6H5NH2)2.H2SO4 Formula Weight: 284.34 Odour: Odourless Solubility: Soluble in water Beilstein Reference: 3729533 UN No.: 2811 MDL No.: MFCD00013110
Formel: C₁₂H₁₆N₂O₄S
Molecular Weight: 284.34 g/mol
Dichte: 1.377
Lagertemperatur: Raumtemperatur
MDL: MFCD00013110
CAS-Nummer: 542-16-5
EINECS: 208-805-0
UN: 2811
ADR: 6.1 III