2-Nitrophenyl-phenylsulfid

Lieferant: Thermo Scientific

BTB15088.250 BTB15088.1
MAYBBTB15088.250EA 0 EUR
MAYBBTB15088.250 MAYBBTB15088.1
2-Nitrophenyl-phenylsulfid
2-Nitrophenyl-phenylsulfid
Formel: C₁₂H₉NO₂S
CAS-Nummer: 4171-83-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR