4-(trans-2-Carboxyvinyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (E)-3-(4-Boronophenyl)acrylic acid

BB-2176-1G BB-2176-5G
COBBBB-2176-1GEA 141 EUR
COBBBB-2176-1G COBBBB-2176-5G
4-(trans-2-Carboxyvinyl)phenylboronsäure
4-(trans-2-Carboxyvinyl)phenylboronsäure
Formel: C₉H₉BO₄
Molecular Weight: 191,9764 g/mol
MDL: MFCD01075704
CAS-Nummer: 159896-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR