4-(trans-2-Carboxyvinyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (E)-3-(4-Boronophenyl)acrylic acid

720569-250MG 720569-1G
SIAL720569-250MGEA 71.8 EUR
SIAL720569-250MG SIAL720569-1G
4-(trans-2-Carboxyvinyl)phenylboronsäure, Sigma-Aldrich®
4-(trans-2-Carboxyvinyl)phenylboronsäure
Formel: C₉H₉BO₄
Molecular Weight: 191,9764 g/mol
MDL: MFCD01075704
CAS-Nummer: 159896-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR