2-(Carboxymethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetic acid

PN-2323-1G PN-2323-250M
COBBPN-2323-1GEA 0 EUR
COBBPN-2323-1G COBBPN-2323-250M
2-(Carboxymethyl)phenylboronsäurepinakolester
2-(Carboxymethyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₉BO₄
MDL: MFCD11504961
CAS-Nummer: 1072945-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR