3-(2-Carboxyethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-(2-Carboxyethyl)benzeneboronic acid, 3-(3-Boronophenyl)propanoic acid
BB-2067-1G
BB-2067-5G
COBBBB-2067-1GEA
0
EUR
COBBBB-2067-1G
COBBBB-2067-5G
3-(2-Carboxyethyl)phenylboronsäure
3-(2-Carboxyethyl)phenylboronsäure
Formel:
C₉H₁₁BO₄ Molecular Weight: 193.99 g/mol |
MDL:
MFCD04115649 CAS-Nummer: 693803-17-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...