4-(2-Carboxyethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(2-Carboxyethyl)benzeneboronic acid

BB-2055-1G BB-2055-5G
COBBBB-2055-1GEA 143 EUR
COBBBB-2055-1G COBBBB-2055-5G
4-(2-Carboxyethyl)phenylboronsäure
4-(2-Carboxyethyl)phenylboronsäure
Formel: C₉H₁₁BO₄
Molecular Weight: 193,99 g/mol
MDL: MFCD01318119
CAS-Nummer: 166316-48-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR