3-(Aminomethyl)phenylboronsäure Hydrochlorid 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: (3-(Aminomethyl)phenyl)boronic acid hydrochloride, 3-(Aminomethyl)benzeneboronic acid hydrochloride

H53284.MD H53284.03
H53284.MDEA 52 EUR
H53284.MD H53284.03
3-(Aminomethyl)phenylboronsäure Hydrochlorid 96%
3-(Aminomethyl)phenylboronsäure Hydrochlorid

Purity: 96% CAS No.: 352525-94-1 Molecular Formula: C7H10BNO2.HCl Formula Weight: 187.43 Melting Point: 248-250° MDL No.: MFCD02093050

Formel: C₇H₁₁BClNO₂
Molecular Weight: 187,43 g/mol
Schmelzpunkt: 248…250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093050
CAS-Nummer: 352525-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR