3-(Aminomethyl)phenylboronsäure Hydrochlorid
Lieferant: COMBI-BLOCKS
Synonyme:
(3-(Aminomethyl)phenyl)boronic acid hydrochloride, 3-(Aminomethyl)benzeneboronic acid hydrochloride
BB-2492-5G
BB-2492-25G
BB-2492-1G
COBBBB-2492-5GEA
0
EUR
COBBBB-2492-5G
COBBBB-2492-25G
COBBBB-2492-1G
3-(Aminomethyl)phenylboronsäure Hydrochlorid
3-(Aminomethyl)phenylboronsäure Hydrochlorid
Formel:
C₇H₁₁BClNO₂ |
MDL:
MFCD02093050 CAS-Nummer: 352525-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...