4-(Aminomethyl)phenylboronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Aminomethylphenylboronic acid hydrochloride, 4-(Aminomethyl)benzeneboronic acid hydrochloride

BB-2443-25G BB-2443-1G BB-2443-5G
COBBBB-2443-25GEA 1030 EUR
COBBBB-2443-25G COBBBB-2443-1G COBBBB-2443-5G
4-(Aminomethyl)phenylboronsäure Hydrochlorid
4-(Aminomethyl)phenylboronsäure Hydrochlorid
Formel: C₇H₁₁BClNO₂
Molecular Weight: 187,43 g/mol
MDL: MFCD01632199
CAS-Nummer: 75705-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR