4-(Aminomethyl)phenylboronsäure Hydrochlorid

Lieferant: FLUOROCHEM

Synonyme: 4-Aminomethylphenylboronic acid hydrochloride, 4-(Aminomethyl)benzeneboronic acid hydrochloride

011014-5G 011014-1G
FLUO011014-5GEA 477 EUR
FLUO011014-5G FLUO011014-1G
4-(Aminomethyl)phenylboronsäure Hydrochlorid
4-(Aminomethyl)phenylboronsäure Hydrochlorid
Formel: C₇H₁₁BClNO₂
Molecular Weight: 187.43 g/mol
MDL: MFCD01632199
CAS-Nummer: 75705-21-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR