5'-Brom-2'-hydroxy-3'-nitroacetophenon
Lieferant: CHEMPUR
Synonyme:
1-(5-Bromo-2-hydroxy-3-nitrophenyl)ethanone
FL12081.POR
012081-25G
012081-5G
CHMPFL12081.POREA
0
EUR
CHMPFL12081.POR
FLUO012081-25G
FLUO012081-5G
5'-Brom-2'-hydroxy-3'-nitroacetophenon
5'-Brom-2'-hydroxy-3'-nitroacetophenon
Formel:
C₈H₆BrNO₄ Molecular Weight: 260.05 g/mol |
MDL:
MFCD01631134 CAS-Nummer: 70978-54-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...