2-Brom-4'-(trifluormethoxy)acetophenon
Lieferant: FLUOROCHEM
Synonyme:
2-Bromo-1-(4-(trifluoromethoxy)phenyl)ethanone, 4-(Trifluoromethoxy)phenacyl bromide
007562-1G
007562-5G
007562-10G
FLUO007562-1GEA
31.1
EUR
FLUO007562-1G
FLUO007562-5G
FLUO007562-10G
2-Brom-4'-(trifluormethoxy)acetophenon
2-Brom-4'-(trifluormethoxy)acetophenon
Formel:
C₉H₆BrF₃O₂ Molecular Weight: 283.04 g/mol |
MDL:
MFCD00052333 CAS-Nummer: 103962-10-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...