2-Brom-4'-(trifluormethoxy)acetophenon

Lieferant: Thermo Scientific

Synonyme: 2-Bromo-1-(4-(trifluoromethoxy)phenyl)ethanone, 4-(Trifluoromethoxy)phenacyl bromide

TL00407 TL00407.0001 TL00407.0003 TL00407.250 TL00407DA
MAYBTL00407EA 0 EUR
MAYBTL00407 MAYBTL00407.0001 MAYBTL00407.0003 MAYBTL00407.250 MAYBTL00407DA
2-Brom-4'-(trifluormethoxy)acetophenon
2-Brom-4'-(trifluormethoxy)acetophenon
Formel: C₉H₆BrF₃O₂
Molecular Weight: 283.04 g/mol
MDL: MFCD00052333
CAS-Nummer: 103962-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR