5-Brom-2-fluorbenzylamin Hydrochlorid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(5-Bromo-2-fluorophenyl)methanamine hydrochloride
B25176.06
B25176.14
B25176.06EA
40.4
EUR
B25176.06
B25176.14
5-Brom-2-fluorbenzylamin Hydrochlorid 97%
5-Brom-2-fluorbenzylamin Hydrochlorid
Purity: 97%
CAS No.: 202865-69-8
Molecular Formula: C7H7BrFN.HCl
Formula Weight: 240.50
Melting Point: 221-226°
MDL No.: MFCD00143426
Formel:
C₇H₈BrClFN Molecular Weight: 240.50 g/mol Schmelzpunkt: 221…226 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143426 CAS-Nummer: 202865-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...