5-Brom-2-fluorbenzylamin Hydrochlorid

Lieferant: FLUOROCHEM

Synonyme: (5-Bromo-2-fluorophenyl)methanamine hydrochloride

008871-1G 008871-5G
FLUO008871-1GEA 45.9 EUR
FLUO008871-1G FLUO008871-5G
5-Brom-2-fluorbenzylamin Hydrochlorid
5-Brom-2-fluorbenzylamin Hydrochlorid
Formel: C₇H₈BrClFN
Molecular Weight: 240.50 g/mol
MDL: MFCD00143426
CAS-Nummer: 202865-69-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR