4-Brom-2-fluorbenzylamin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: (4-Bromo-2-fluorophenyl)methanamine hydrochloride

FL13225.POR 013225-5G
CHMPFL13225.POREA 0 EUR
CHMPFL13225.POR FLUO013225-5G
4-Brom-2-fluorbenzylamin Hydrochlorid
4-Brom-2-fluorbenzylamin Hydrochlorid
Formel: C₇H₇BrFN·HCl
Molecular Weight: 240.50 g/mol
MDL: MFCD00143266
CAS-Nummer: 147181-08-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR