4-Chlor-3-(trifluormethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(4-Chloro-3-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-3502-5G PN-3502-1G
COBBPN-3502-5GEA 130 EUR
COBBPN-3502-5G COBBPN-3502-1G
4-Chlor-3-(trifluormethyl)phenylboronsäurepinakolester
4-Chlor-3-(trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₃H₁₅BClF₃O₂
Molecular Weight: 306,516 g/mol
MDL: MFCD06657823
CAS-Nummer: 445303-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR