3-Chlor-N,2-dimethylanilin

Lieferant: COMBI-BLOCKS

AN-1466-5G AN-1466-1G
COBBAN-1466-5GEA 1100 EUR
COBBAN-1466-5G COBBAN-1466-1G
3-Chlor-N,2-dimethylanilin
3-Chlor-N,2-dimethylanilin
Formel: C₈H₁₀ClN
MDL: MFCD10687534
CAS-Nummer: 41456-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR