3-Chlor-N,2-dimethylanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

AN-1246-5G AN-1246-1G
COBBAN-1246-5GEA 451 EUR
COBBAN-1246-5G COBBAN-1246-1G
3-Chlor-N,2-dimethylanilin Hydrochlorid
3-Chlor-N,2-dimethylanilin Hydrochlorid
Formel: C₈H₁₁Cl₂N
MDL: MFCD09878314
CAS-Nummer: 957062-82-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR