3-(Benzylamino)propionitril ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-(Benzylamino)propanenitrile
41949.06
41949.14
41949.22
41949.06EA
28.1
EUR
41949.06
41949.14
41949.22
3-(Benzylamino)propionitril ≥97%
3-(Benzylamino)propionitril
Purity: 97+%
CAS No.: 706-03-6
Molecular Formula: C6H5CH2NHCH2CH2CN
Formula Weight: 160.22
Flash Point:
Formel:
C₁₀H₁₂N₂ Molecular Weight: 160.22 g/mol Dichte: 1.024 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001955 CAS-Nummer: 706-03-6 EINECS: 211-890-7 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...