3-(Benzylamino)propionitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Benzylamino)propanenitrile

185167-100G 185167-25G
SIAL185167-100GEA 198 EUR
SIAL185167-100G SIAL185167-25G
3-(Benzylamino)propionitril, Sigma-Aldrich®
3-(Benzylamino)propionitril
Formel: C₁₀H₁₂N₂
Molecular Weight: 160,22 g/mol
MDL: MFCD00001955
CAS-Nummer: 706-03-6
EINECS: 211-890-7
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR