2'-Amino-4',5'-dimethoxyacetophenon 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(2-Amino-4,5-dimethoxyphenyl)ethanone

L10605.06 L10605.14
L10605.06EA 110 EUR
L10605.06 L10605.14
2'-Amino-4',5'-dimethoxyacetophenon 98%
2'-Amino-4',5'-dimethoxyacetophenon

Purity: 98% CAS No.: 4101-30-8 Molecular Formula: C10H13NO3 Formula Weight: 195.22 Melting Point: 104-108° Beilstein Reference: 391380

Formel: C₁₀H₁₃NO₃
Schmelzpunkt: 104…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016646
CAS-Nummer: 4101-30-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR