2-(p-Toluolsulfonyl)acetophenon
Lieferant: FLUOROCHEM
003788-5G
003788-25G
FLUO003788-5GEA
0
EUR
FLUO003788-5G
FLUO003788-25G
2-(p-Toluolsulfonyl)acetophenon
2-(p-Toluolsulfonyl)acetophenon
Formel:
C₁₅H₁₄O₃S |
MDL:
MFCD00026002 CAS-Nummer: 31378-03-7 EINECS: 250-605-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...