4-Methoxy-2-(trifluormethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(4-Methoxy-2-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-8402-1G PN-8402-5G
COBBPN-8402-1GEA 204 EUR
COBBPN-8402-1G COBBPN-8402-5G
4-Methoxy-2-(trifluormethyl)phenylboronsäurepinakolester
4-Methoxy-2-(trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₈BF₃O₃
Molecular Weight: 302,100 g/mol
MDL: MFCD09746197
CAS-Nummer: 1218790-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR