2-Formyl-4-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Formyl-4-(trifluoromethyl)phenyl)boronic acid

BB-6083-5G BB-6083-1G
COBBBB-6083-5GEA 1700 EUR
COBBBB-6083-5G COBBBB-6083-1G
2-Formyl-4-(trifluormethyl)phenylboronsäure
2-Formyl-4-(trifluormethyl)phenylboronsäure
Formel: C₈H₆BF₃O₃
Molecular Weight: 217.9400 g/mol
MDL: MFCD13195656
CAS-Nummer: 1217501-35-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR