4-Formyl-3-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Formyl-3-(trifluoromethyl)benzeneboronic acid

BB-8577-1G BB-8577-250M
COBBBB-8577-1GEA 514 EUR
COBBBB-8577-1G COBBBB-8577-250M
4-Formyl-3-(trifluormethyl)phenylboronsäure
4-Formyl-3-(trifluormethyl)phenylboronsäure
Formel: C₈H₆BF₃O₃
Molecular Weight: 217.94 g/mol
MDL: MFCD09037498
CAS-Nummer: 1072944-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR